missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
NFkB2/NFkB p100 Monoclonal specifically detects NFkB2/NFkB p100 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | NFkB2/NFkB p100 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | CL8266 |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Formulation | PBS, pH 7.2, containing 40% glycerol |
| Host Species | Mouse |
| Immunogen | This antibody was developed against a synthetic peptide corresponding to amino acids: LKRKRGGDVSDSKQFTYYP |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?