missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFkB2/NFkB p100 Antibody (CL8266) [HRP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-88914H
This item is not returnable.
View return policy
Description
NFkB2/NFkB p100 Monoclonal antibody specifically detects NFkB2/NFkB p100 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| NFkB2/NFkB p100 | |
| Monoclonal | |
| HRP | |
| CVID10;H2TF1;LYT10;LYT-10;NF-kB2;Nuclear factor NF-kappa-B p100 subunit;p100;p49/p100;p52 | |
| This antibody was developed using a synthetic peptide derived from Q00653, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Apoptosis, B Cell Development and Differentiation Markers, Cancer, Cell Biology, Immunology, Signal Transduction | |
| 4791 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL8266 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction