missing translation for 'onlineSavingsMsg'
Learn More

NFkB2/NFkB p100 Antibody (291319) [DyLight 594], Novus Biologicals™

Product Code. 30733066 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30733066 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733066 Supplier Novus Biologicals Supplier No. FAB28881M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NFkB2/NFkB p100 Monoclonal antibody specifically detects NFkB2/NFkB p100 in Human samples. It is validated for Western Blot,ChIP assay (ChIP),Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFkB2/NFkB p100
Applications Western Blot, ChIP Assay, Immunocytochemistry
Classification Monoclonal
Clone 291319
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias CVID10;H2TF1;LYT10;LYT-10;NF-kB2;Nuclear factor NF-kappa-B p100 subunit;p100;p49/p100;p52
Host Species Mouse
Immunogen E. coli-derived recombinant human NF kappa B2, Met1-Asn447, Accession # Q00653
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, B Cell Development and Differentiation Markers, Cancer, Cell Biology, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4791
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.