missing translation for 'onlineSavingsMsg'
Learn More

NFkB1/NFkB p105 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18384444 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18384444 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18384444 Supplier Novus Biologicals Supplier No. NBP305867AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NFkB1/NFkB p105 Polyclonal specifically detects NFkB1/NFkB p105 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFkB1/NFkB p105
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias DNA-binding factor KBF1, EBP-1, NF-kappa-B, NF-kB1, NFKB1, NFKB-p105, Nuclear factor NF-kappa-B p105 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 1, p105
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human NFkB1/NFkB p105.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4790
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.