missing translation for 'onlineSavingsMsg'
Learn More

NFkB p105/p50 Antibody, Novus Biologicals™

Product Code. 18404172 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18404172 0.1 mL 0.1mL
18437671 25 μL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18404172 Supplier Novus Biologicals Supplier No. NBP187758

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NFkB p105/p50 Polyclonal antibody specifically detects NFkB p105/p50 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFkB p105/p50
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:200 - 1:500, Knockdown Validated
Formulation PBS (pH 7.2) and 40% Glycerol
Gene Alias DKFZp686C01211, DNA binding factor KBF1, DNA-binding factor KBF1, EBP-1, KBF1, NF-kappaB, NF-kappa-B, NF-kappabeta, NFKB-p105, NFKB-p50, nuclear factor kappa-B DNA binding subunit, nuclear factor NF-kappa-B p105 subunit, nuclear factor NF-kappa-B p50 subunit, nuclear factor of kappa light polypeptide gene enhancer in B-cells 1MGC54151, p105, p50
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: GTGSTGPGYSFPHYGFPTYGGITFHPGTTKSNAGMKHGTMDTESKKDPEGCDKSDDKNTVNLFGKVIETTEQDQEPSEATVGNGEVTLTYATGTKEESAGVQDNLFLEKAMQLAKRHANALFDYAVTGDVKMLLAVQRHLTAV
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Diabetes Research, DNA Repair, Neurodegeneration, Neuroscience, Nucleotide Excision Repair, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 4790
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.