missing translation for 'onlineSavingsMsg'
Learn More

NFkB p105/p50 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£164.00 - £442.00

Specifications

Antigen NFkB p105/p50
Dilution Western Blot 1:100 - 1:500, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30232575
View Documents
Novus Biologicals
NBP3-38305-20ul
20 μL
£164.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30227036
View Documents
Novus Biologicals
NBP3-38305-100ul
100 μL
£442.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NFkB p105/p50 Polyclonal antibody specifically detects NFkB p105/p50 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications

Specifications

NFkB p105/p50
ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Unconjugated
Rabbit
Apoptosis, Cancer, Cell Biology, Diabetes Research, DNA Repair, Neurodegeneration, Neuroscience, Nucleotide Excision Repair, Phospho Specific, Signal Transduction, Transcription Factors and Regulators
PBS (pH 7.3), 50% glycerol
4790
IgG
Affinity purified
Western Blot 1:100 - 1:500, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Polyclonal
Purified
RUO
Human, Mouse, Rat
DKFZp686C01211, DNA binding factor KBF1, DNA-binding factor KBF1, EBP-1, KBF1, NF-kappaB, NF-kappa-B, NF-kappabeta, NFKB-p105, NFKB-p50, nuclear factor kappa-B DNA binding subunit, nuclear factor NF-kappa-B p105 subunit, nuclear factor NF-kappa-B p50 subunit, nuclear factor of kappa light polypeptide gene enhancer in B-cells 1MGC54151, p105, p50
A synthetic peptide corresponding to a sequence within amino acids 41-365 of human NFkB p105/p50 (NP_001158884.1).,, Sequence:, LAGCLLLEGDAHVDSTTYDGTTPLHIAAGRGSTRLAALLKAAGADPLVENFEPLYDLDDSWENAGEDEGVVPGTTPLDMATSWQVFDILNGKPYEPEFTSD
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.