missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFkB p105/p50 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35166-100ul
This item is not returnable.
View return policy
Description
NFkB p105/p50 Polyclonal antibody specifically detects NFkB p105/p50 in Human samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| NFkB p105/p50 | |
| Polyclonal | |
| Western Blot 1:100 - 1:500, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:20 - 1:100, Immunoprecipitation 0.5ug - 4ug antibody for 200ug - 400ug extracts of whole cells, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| DKFZp686C01211, DNA binding factor KBF1, DNA-binding factor KBF1, EBP-1, KBF1, NF-kappaB, NF-kappa-B, NF-kappabeta, NFKB-p105, NFKB-p50, nuclear factor kappa-B DNA binding subunit, nuclear factor NF-kappa-B p105 subunit, nuclear factor NF-kappa-B p50 subunit, nuclear factor of kappa light polypeptide gene enhancer in B-cells 1MGC54151, p105, p50 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 740-964 of NFkB p105/p50 (NP_001158884.1).,, Sequence:, AAGADPLVENFEPLYDLDDSWENAGEDEGVVPGTTPLDMATSWQVFDILNGKPYEPEFTSDDLLAQGDMKQLAEDVKLQLYKLLEIPDPDKNWATLAQKLGLGILNNAFRLSPAPSKTLMDNYEVSGGTVRELVEALRQMGYTEAIEVIQAASSPVKTTSQAHSLPLSPASTRQQIDELRDSDSVCDSGVETSFRKLSFTESLTSGASLLTLNKMPHDYGQEGPL | |
| 100 μL | |
| Apoptosis, Cancer, Cell Biology, Diabetes Research, DNA Repair, Neurodegeneration, Neuroscience, Nucleotide Excision Repair, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
| 4790 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction