missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFkB p100/p52 Rabbit anti-Human, Mouse, Rat, Clone: S04-4D6, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-14959-100UL
This item is not returnable.
View return policy
Description
NFkB p100/p52 Monoclonal antibody specifically detects NFkB p100/p52 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| NFkB p100/p52 | |
| Monoclonal | |
| Unconjugated | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
| Rabbit | |
| Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| S04-4D6 | |
| Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:10-1:500 | |
| DNA-binding factor KBF2, H2TF1, Lymphocyte translocation chromosome 10 protein, Lyt10, LYT10LYT-10, nuclear factor NF-kappa-B p100 subunit, nuclear factor of kappa light chain gene enhancer in B-cells 2, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 2, nuclear factor of kappa light polypeptide gene enhancer in B-cells 2 (p49/p100), Oncogene Lyt-10, p52 | |
| Recombinant protein of human NFkB p100/p52 (Uniprot#: Q00653) | |
| 100 μg | |
| Apoptosis, Cancer, Cell Biology, Neurodegeneration, Neuroscience, Signal Transduction, Transcription Factors and Regulators | |
| 4791 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction