missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NFIL3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106504
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total NFIL3.
Expression of interleukin-3 (IL3; MIM 147740) is restricted to activated T cells, natural killer (NK) cells, and mast cell lines. Transcription initiation depends on the activating capacity of specific protein factors, such as NFIL3, that bind to regulatory regions of the gene, usually upstream of the transcription start site (Zhang et al., 1995 [PubMed 7565758]).
Specifications
| NFIL3 | |
| Polyclonal | |
| Unconjugated | |
| NFIL3 | |
| AV225605; E4 promoter binding-protein 4; E4 promoter-binding protein 4; E4BP4; Embryo implantation-related NFIL3/E4BP4-like transcription factor; IL3BP1; Interleukin-3 promoter transcriptional activator; interleukin-3-binding protein 1; NF IL3A; NFIL3; NFIL3A; NF-IL3A; nuclear factor interleukin 3 regulated protein; nuclear factor interleukin-3-regulated protein; nuclear factor, interleukin 3 regulated; nuclear factor, interleukin 3, regulated; OTTHUMP00000021633; transcriptional activator NF-IL3A | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 114519, 18030, 4783 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O08750, Q16649, Q6IMZ0 | |
| NFIL3 | |
| A synthesized peptide derived from human NFIL3(Accession Q16649), corresponding to amino acid residues E429-G462. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction