missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFIL3 Mouse anti-Human, Porcine, Clone: 2F4B4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66368-1-IG-20UL
This item is not returnable.
View return policy
Description
MCM2 (Mini chromosome maintece protein 2) is involved in regulating DNA replication. MCM2 (also called CDCL1, mitotin and BM28), is a human nuclear protein that is crucial in the cell cycle, being involved in the onset of DNA replication and cell division. It is similar to members of the family of early S-phase proteins. Mincheva et al. (1994) mapped the gene to 3q21. From its localization, CDCL1 became a candidate for an oncogene affected by chromosomal breaks in acute myeloid leukemia (AML).Specifications
| NFIL3 | |
| Monoclonal | |
| 0.93 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q16649 | |
| NFIL3 | |
| NFIL3 Fusion Protein Ag2380 | |
| 20 μL | |
| Primary | |
| Pig, Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 2F4B4 | |
| Unconjugated | |
| NFIL3 | |
| E4 promoter binding protein 4, E4BP4, IL3BP1, NF IL3A, NFIL3, NFIL3A | |
| Mouse | |
| Protein G | |
| RUO | |
| 100153822, 4783 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction