missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFIL3 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-41077AF700
This item is not returnable.
View return policy
Description
NFIL3 Polyclonal antibody specifically detects NFIL3 in Human samples. It is validated for Western Blot
Specifications
| NFIL3 | |
| Polyclonal | |
| Western Blot | |
| E4BP4IL3BP1, Interleukin-3 promoter transcriptional activator, Interleukin-3-binding protein 1, NF-IL3A, NFIL3AE4 promoter-binding protein 4, nuclear factor interleukin 3 regulated protein, nuclear factor interleukin-3-regulated protein, nuclear factor, interleukin 3 regulated, Transcriptional activator NF-IL3A | |
| The immunogen for this product maps to a region between residue 412 and 462 of human E4 promoter binding-protein 4 using the numbering given in entry NP_005375.2 (GeneID 4783). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 4783 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction