missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFIL3 Antibody (1218A), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB8888CL1
This item is not returnable.
View return policy
Description
NFIL3 Monoclonal antibody specifically detects NFIL3 in Mouse samples. It is validated for Flow Cytometry (Intracellular Staining)
Specifications
| NFIL3 | |
| Monoclonal | |
| CoraFluor 1 | |
| E4BP4IL3BP1, Interleukin-3 promoter transcriptional activator, Interleukin-3-binding protein 1, NF-IL3A, NFIL3AE4 promoter-binding protein 4, nuclear factor interleukin 3 regulated protein, nuclear factor interleukin-3-regulated protein, nuclear factor, interleukin 3 regulated, Transcriptional activator NF-IL3A | |
| E. coli-derived recombinant mouse NFIL3/E4BP4, Lys140-Arg462, Accession # O08750 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry (Intracellular Staining) | |
| 1218A | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| 4783 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction