missing translation for 'onlineSavingsMsg'
Learn More

NFIC Antibody, DyLight 680, Novus Biologicals™

Product Code. 18416604 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18416604 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18416604 Supplier Novus Biologicals Supplier No. NBP171879FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NFIC Polyclonal antibody specifically detects NFIC in Human samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFIC
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate DyLight 680
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias CCAAT-binding transcription factor, CTF5, CTFCCAAT-box-binding transcription factor, MGC20153, NF1-C, NFI, NF-I, NF-I/C, NFI-C, nuclear factor 1 C-type, Nuclear factor 1/C, Nuclear factor I/C, nuclear factor I/C (CCAAT-binding transcription factor), TGGCA-binding protein
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 250 and 300 of human Nuclear Factor I/C using the numbering given in entry EAW69324.1 (GeneID 4782).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 4782
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.