missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFIB Antibody (S03-0E5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19716-50ul
This item is not returnable.
View return policy
Description
NFIB Monoclonal antibody specifically detects NFIB in Human, Rat samples. It is validated for Western Blot
Specifications
| NFIB | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot | |
| S03-0E5 | |
| Unconjugated | |
| CCAAT-box-binding transcription factor, CTF, HMGIC/NFIB, NF-I/B, NFI-B, NFIB2, NFIB3, NFI-RED, nuclear factor 1 B-type, Nuclear factor 1/B, nuclear factor I/BNF1-B, TGGCA-binding protein | |
| A synthetic peptide of human NFIB (Uniprot # O00712) | |
| 50 μg | |
| Cell Cycle and Replication | |
| 4781 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction