missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFIA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11750-1-AP-150UL
This item is not returnable.
View return policy
Description
11750-1-AP is a rabbit polyclonal antibody raised against N-terminal of human original NFIA.
NF-1, also designated CTF, consists of a family of CCAAT box binding proteins that stimulate DNA replication and activate transcription. Analysis of human NF-1 messenger RNA has revealed two forms of the NF-1 protein arising from an alternate splicing of a single NF-1 gene. NF-1 binds its consensus DNA element as a homodimer via an amino-terminal DNA binding domain, and activates transcription through a putatively novel, proline-rich, carboxy terminal transactivation domain. The NF-1 protein has been shown to recognize and bind the adenovirus type 2 promoter and activate transcription of herpes simplex virus thymidine kinase genes. The NF-1 consensus element has been found in the upstream promoter region of myriad eukaryotic genes, including that of Ha-Ras, alpha-globin, HSP 70, GRP 78, Histone H1, myelin basic protein and in the Xenopus laevis vitellogenin gene promoter.Specifications
| NFIA | |
| Polyclonal | |
| Unconjugated | |
| NFIA | |
| CTF, KIAA1439, NF I/A, NF1 A, NFI A, NFI L, NFIA, Nuclear factor 1 A type, Nuclear factor 1/A, nuclear factor I/A, TGGCA binding protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18027, 25492, 4774 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P09414, Q02780, Q12857 | |
| Nfia | |
| NFIA Fusion Protein Ag2346 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction