missing translation for 'onlineSavingsMsg'
Learn More

NFIA Antibody (PCRP-NFIA-2C6), Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18351757 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18351757 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18351757 Supplier Novus Biologicals Supplier No. NBP314016AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NFIA Monoclonal antibody specifically detects NFIA in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFIA
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PCRP-NFIA-2C6
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias CCAAT-box-binding transcription factor, CTF, DKFZp686J23256, FLJ39164, KIAA1439DKFZp434L0422, NF1-A, NF-I/A, NFI-A, NFI-L, nuclear factor 1 A-type, Nuclear factor 1/A, nuclear factor I/ATGGCA-binding protein
Host Species Mouse
Immunogen Recombinant fragment of human NFIA protein corresponding to protein domain (Uniprot: Q15784)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 4774
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2c κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.