missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NFIC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5107192
This item is not returnable.
View return policy
Description
NFIC Polyclonal Antibody for Western Blot
Recognizes and binds the palindromic sequence 5'-TTGGCNNNNNGCCAA-3' present in viral and cellular promoters and in the origin of replication of adenovirus type 2. These proteins are individually capable of activating transcription and replication.
Specifications
| NFIC | |
| Polyclonal | |
| Unconjugated | |
| NFIC | |
| 1110019L22Rik; 1500041O16Rik; AA589446; AI746521; CCAAT-box-binding transcription factor; CTF; CTF5; KIAA1439; NF1-A; NF1-B; NF1-C; NF1-X; NFI; NF-I; NF-I/A; NF-I/B; NF-I/C; NF-I/X; NFIA; NFI-A; NFIB; NFI-B; Nfic; NFI-C; NFIX; NFI-X; Nuclear factor 1 C-type; Nuclear factor 1/A; Nuclear factor 1/B; nuclear factor 1/C; Nuclear factor 1/X; nuclear factor 1-C2; nuclear factor I C; Nuclear factor I/A; Nuclear factor I/B; nuclear factor I/C; nuclear factor I/C (CCAAT-binding transcription factor); Nuclear factor I/X; olfactory epithelium nuclear factor I-C; TGGCA-binding protein | |
| Rabbit | |
| Protein A | |
| RUO | |
| 4782 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| P08651 | |
| NFIC | |
| Synthetic peptide directed towards the C-terminal region of human NFIC. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction