missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFE2L1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17062-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is a rabbit polyclonal antibody raised against residues near the C terminus of human ESRRG.
This gene encodes a protein that is involved in globin gene expression in erythrocytes. Confusion has occurred in bibliographic databases due to the shared symbol of NRF1 for this gene, NFE2L1, and for ′nuclear respiratory factor 1′ which has an official symbol of NRF1.Specifications
| NFE2L1 | |
| Polyclonal | |
| Unconjugated | |
| NFE2L1 | |
| HBZ17, LCR F1, Locus control region factor 1, NF E2 related factor 1, NFE2 related factor 1, NFE2L1, NRF1, TCF 11, TCF11, Transcription factor 11, Transcription factor HBZ17, Transcription factor LCR F1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18023, 4779 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q14494, Q61985 | |
| Nfe2l1 | |
| NFE2L1 Fusion Protein Ag10832 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction