missing translation for 'onlineSavingsMsg'
Learn More

NFATC3/NFAT4 Antibody, DyLight 488, Novus Biologicals™

Product Code. 18413074 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18413074 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18413074 Supplier Novus Biologicals Supplier No. NBP204025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NFATC3/NFAT4 Polyclonal antibody specifically detects NFATC3/NFAT4 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFATC3/NFAT4
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias NF-AT4, NFAT4nuclear factor of activated T-cells, cytoplasmic 3, NFATc3, NF-ATc3, NFATx, nuclear factor of activated T-cells c3 isoform IE-Xa, nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 3, T cell transcription factor NFAT4, T-cell transcription factor NFAT4
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 100 and 150 of human Nuclear Factor of Activated T-cells, Cytoplasmic, Calcineurin-dependent 3 using the numbering given in entry NP_775188.1 (GeneID 4775).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 4775
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.