Learn More
NFATC3 Mouse anti-Human, Unconjugated, Clone: 1C1H8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66924-1-IG-20UL
This item is not returnable.
View return policy
Description
The NFATC3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NFATC3. This antibody recognizes human antigen. The NFATC3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| NFATC3 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot | |
| 1C1H8 | |
| Unconjugated | |
| 20 μL | |
| 4775 | |
| IgG1 |