Learn More
Invitrogen™ NFATC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA14198
Description
PA1-4198 detects NFAT2 in human samples. NFATc2 is also known as NFAT1 or NFATp. PA1-4198 has been successfully used in Western blot procedures. The PA1-4198 immunogen is a synthetic peptide corresponding to residues A(269) S P Q R S R S P S P Q P(281) of human NFATc2.
The nuclear factor of activated T-cells (NFAT) transcription complex is required for the expression of a group of proteins that collectively regulate the immune response. Four NFAT proteins, encoded on separate genes and expressed as several splice variants, have been described: NFAT1 (also known as NFATp or NFATc2), NFAT2 (NFATc or NFATc1), NFAT3, and NFAT4 (NFATx or NFATc3). These proteins show a low level of sequence similarity with the Dorsal/Rel/NFkB family of transcription factors. Another NFAT-related protein termed NFAT5 differs from isoforms 1-4 in that it lacks many of the Fos/Jun contact sites observed in its predecessors and its subcellular localization is not calcineurin-dependent.
Specifications
| NFATC2 | |
| Polyclonal | |
| Unconjugated | |
| NFATC2 | |
| AI607462; IB-IXL; KIAA0611; NFAT pre-existing s; NFAT pre-existing subunit; NFAT transcription complex, preexisting component; Nfat1; NFAT1-D; Nfatc2; NF-ATc2; Nfatp; NF-ATp; nuclear factor of activated T cells, cytoplasmic, calcineurin dependent 2; nuclear factor of activated T-cells 2; nuclear factor of activated T-cells, cytoplasmic 2; nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 2; nuclear factor of activated T-cells, preexisting component; OTTHUMP00000031291; preexisting nuclear factor of activated T-cells 2; T cell transcription factor NFAT1; T-cell transcription factor NFAT1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4773 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q13469 | |
| NFATC2 | |
| Synthetic peptide corresponding to residues A(269) S P Q R S R S P S P Q P(281) of human NFATc2. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.