missing translation for 'onlineSavingsMsg'
Learn More

NFATC2/NFAT1 Antibody (2A4), Novus Biologicals™

Product Code. 18321268 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18321268 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18321268 Supplier Novus Biologicals Supplier No. H00004773M01100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NFATC2/NFAT1 Monoclonal antibody specifically detects NFATC2/NFAT1 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFATC2/NFAT1
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 2A4
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_036472
Gene Alias NFAT pre-existing subunit, NFAT transcription complex, preexisting component, NFAT1nuclear factor of activated T-cells, cytoplasmic 2, NFATc2, NF-ATc2, NFATp, NF-ATp, NFATPnuclear factor of activated T-cells, preexisting component, nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 2, preexisting nuclear factor of activated T-cells 2, T cell transcription factor NFAT1, T-cell transcription factor NFAT1
Host Species Mouse
Immunogen NFATC2 (NP_036472, 812 a.a. ~ 921 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. HYSPTNQQLRCGSHQEFQHIMYCENFAPGTTRPGPPPVSQGQRLSPGSYPTVIQQQNATSQRAAKNGPPVSDQKEVLPAGVTIKQEQNLDQTYLDDELIDTRLSWIQNIL
Purification Method IgG purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Transcription Factors and Regulators, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 4773
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.