missing translation for 'onlineSavingsMsg'
Learn More

NFAT5 Antibody, Novus Biologicals™

Product Code. 18340149 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18340149 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18340149 Supplier Novus Biologicals Supplier No. H00010725B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

NFAT5 Polyclonal antibody specifically detects NFAT5 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFAT5
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_619727.1
Gene Alias EC 3.6.1, EC 3.6.3.14, KIAA0827glutamine rich protein H65, NF-AT5NFAT-like protein 1, NFATL1, NFATZ, nuclear factor of activated T-cells 5, tonicity-responsive, OREBP, osmotic response element-binding protein, T-cell transcription factor NFAT5, TonE-binding protein, TonEBP, TONEBPnuclear factor of activated T-cells 5, Tonicity-responsive enhancer-binding protein
Host Species Mouse
Immunogen NFAT5 (NP_619727.1, 1 a.a. - 1484 a.a.) full-length human protein. MSQTSGGEAGSPPPAVVAADASSAPSSSSMGGACSSFTTSSSPTIYSTSVTDSKAMQVESCSSAVGVSNRGVSEKQLTSNTVQQHPSTPKRHTVLYISPPPEDLLDNSRMSCQDEGCGLESEQSCSMWMEDSPSNFSNMSTSSYNDNTEVPRKSRKRNPKQRPGVKRRDCEESNMDIFDADSAKAPHYVLSQLTTDNKGNSKAGNGTLENQKGTGVKKSPMLCGQYPVKSEGKELKIVVQPETQHRARYLTEGSRGSVKDRTQQGFPTVKLEGHNEPVVLQVFVGNDSGRVKPHGFYQACRVTGRNTTPCKEVDIEGTTVIEVGLDPSNNMTLAVDCVGILKLRNADVEARIGIAGSKKKSTRARLVFRVNIMRKDGSTLTLQTPSSPILCTQPAGVPEILKKSLHSCSVKGEEEVFLIGKNFLKGTKVIFQENVSDENSWKSEAEIDMELFHQNHLIVKVPPYHDQHITLPVSVGIYVVTNAGRSHDVQPFTYTPDPAAAGALNVNVKKEISSPARPCSFEEAMKAMKTTGCNLDKVNIIPNALMTPLIPSSMIKSEDVTPMEVTAEKRSSTIFKTTKSVGSTQQTLENISNIAGNGSFSSPSSSHLPSENEKQQQIQPKAYNPETLTTIQTQDISQPGTFPAVSASSQLPNSDALLQQATQFQTRETQSREILQSDGTVVNLSQLTEASQQQQQSPLQEQAQTLQQQISSNIFPSPNSVSQLQNTIQQLQAGSFTGSTASGSSGSVDLVQQVLEAQQQLSSVLFSAPDGNENVQEQLSADIFQQVSQIQSGVSPGMFSSTEPTVHTRPDNLLPGRAESVHPQSENTLSNQQQQQQQQQQVMESSAAMVMEMQQSICQAAAQIQSELFPSTASANGNLQQSPVYQQTSHMMSALSTNEDMQMQCELFSSPPAVSGNETSTTTTQQVATPGTTMFQTSSSGDGEETGTQAKQIQNSVFQTMVQMQHSGDNQPQVNLFSSTKSMMSVQNSGTQQQGNGLFQQGNEMMSLQSGNFLQQSSHSQAQLFHPQNPIADAQNLSQETQGSLFHSPNPIVHSQTSTTSSEQMQPPMFHSQSTIAVLQGSSVPQDQQSTNIFLSQSPMNNLQTNTVAQEAFFAAPNSISPLQSTSNSEQQAAFQQQAPISHIQTPMLSQEQAQPPQQGLFQPQVALGSLPPNPMPQSQQGTMFQSQHSIVAMQSNSPSQEQQQQQQQQQQQQQQQQQSILFSNQNTMATMASPKQPPPNMIFNPNQNPMANQEQQNQSIFHQQSNMAPMNQEQQPMQFQSQSTVSSLQNPGPTQSESSQTPLFHSSPQIQLVQGSPSSQEQQVTLFLSPASMSALQTSINQQDMQQSPLYSPQNNMPGIQGATSSPQPQATLFHNTAGGTMNQLQNSPGSSQQTSGMFLFGIQNNCSQLLTSGPATLPDQLMAISQPGQPQNEGQPPVTTLLSQQMPENSPLASSINTNQNIEKIDLLVSLQNQGNNLTGSF
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 10725
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.