missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFAT5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35688-20ul
This item is not returnable.
View return policy
Description
NFAT5 Polyclonal antibody specifically detects NFAT5 in Human samples. It is validated for ELISA,Western Blot
Specifications
| NFAT5 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| EC 3.6.1, EC 3.6.3.14, KIAA0827glutamine rich protein H65, NF-AT5NFAT-like protein 1, NFATL1, NFATZ, nuclear factor of activated T-cells 5, tonicity-responsive, OREBP, osmotic response element-binding protein, T-cell transcription factor NFAT5, TonE-binding protein, TonEBP, TONEBPnuclear factor of activated T-cells 5, Tonicity-responsive enhancer-binding protein | |
| A synthetic peptide corresponding to a sequence within amino acids 1355-1455 of human NFAT5 (NP_619728.2).,, Sequence:, LSPASMSALQTSINQQDMQQSPLYSPQNNMPGIQGATSSPQPQATLFHNTAGGTMNQLQNSPGSSQQTSGMFLFGIQNNCSQLLTSGPATLPDQLMAISQPGQPQNEGQPPVTTLLSQQMPENSPLASSINTNQNIEKIDLLVSLQNQGNNLTGSF | |
| 20 μL | |
| Wnt Signaling Pathway | |
| 10725 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction