missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NFAM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103469
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total NFAM1.
The protein encoded by this gene is a type I membrane receptor that activates cytokine gene promoters such as the IL-13 and TNF-alpha promoters. The encoded protein contains an immunoreceptor tyrosine-based activation motif (ITAM) and is thought to regulate the signaling and development of B-cells. It is also known CNAIP.
Specifications
| NFAM1 | |
| Polyclonal | |
| Unconjugated | |
| NFAM1 | |
| 4921501M20Rik; bK126B4.4; Calcineurin/NFAT-activating ITAM-containing protein; Cnaip; CTA-126B4.4; Nfam1; Nfat activating molecule with ITAM motif 1; NFAT activating protein with ITAM motif 1; NFAT activation molecule 1; NFAT-activating protein with ITAM motif 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 150372, 74039 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8NET5, Q8R4V1 | |
| NFAM1 | |
| A synthesized peptide derived from human NFAM1(Accession Q8NET5), corresponding to amino acid residues L183-C233. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction