missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NF45 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14714-1-AP-150UL
This item is not returnable.
View return policy
Description
Nuclear factor of activated T-cells is a transcription factor required for T-cell expression of the interleukin 2 gene. NFAT binds to a sequence in the interleukin 2 gene enhancer known as the antigen receptor response element 2. In addition, NFAT can bind RNA and is an essential component for encapsidation and protein priming of hepatitis B viral polymerase. NFAT is a heterodimer of 45 kDa and 90 kDa proteins, the smaller of which is the product of this gene. The encoded protein binds strongly to the 90 kDa protein and stimulates its ability to enhance gene expression.Specifications
| NF45 | |
| Polyclonal | |
| Unconjugated | |
| ILF2 | |
| ILF2, NF45, PRO3063 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3608, 67781 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q12905, Q9CXY6 | |
| ILF2 | |
| NF45 Fusion Protein Ag6436 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction