missing translation for 'onlineSavingsMsg'
Learn More

NF2 Recombinant Rabbit Monoclonal Antibody (1H27L4)

Product Code. 15933854
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15933854 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15933854 Supplier Invitrogen Supplier No. 702466

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

NF2 Recombinant Rabbit Monoclonal Antibody (1H27L4)

NF2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

TRUSTED_SUSTAINABILITY

Specifications

Antigen NF2
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 1H27L4
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Nf2
Gene Accession No. P35240, P46662, Q63648
Gene Alias ACN; BANF; Merlin; MGC123124; moesin-ezrin-radixin like; moesin-ezrin-radixin-like protein; moesin-ezrin-radizin-like protein; neurofibromatosis 2; neurofibromin 2; neurofibromin 2 (bilateral acoustic neuroma); neurofibromin 2 (merlin); neurofibromin 2b (merlin); neurofibromin-2; Nf2; Nf-2; NF2a; nf2b; OTTHUMP00000199607; SCH; Schwannomerlin; schwannomin
Gene Symbols Nf2
Host Species Rabbit
Immunogen Protein corresponding to human NF2 [aa331-aa530].
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18016, 25744, 4771
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.