Learn More
NF2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA589329
Description
Positive Samples: A549, A431, HeLa, SKOV3, Mouse brain, Mouse kidney, Mouse heart, Rat brain; Cellular Location: Cell projection, Cytoplasm, Cytoplasmic granule, Cytoplasmic side, Nucleus, Nucleus, Peripheral membrane protein, cytoskeleton, filopodium membrane, perinuclear region, ruffle membrane Immunogen sequence: TKPTYPPMNP IPAPLPPDIP SFNLIGDSLS FDFKDTDMKR LSMEIEKEKV EYMEKSKHLQ EQLNELKTEI EALKLKERET ALDILHNENS DRGGSSKHNT
NF2 encodes a protein, Merlin, that is similar to some members of the ERM family of proteins that are thought to link cytoskeletal components with proteins in the cell membrane. Merlin has been shown to interact with cell-surface proteins, proteins involved in cytoskeletal dynamics and proteins involved in regulating ion transport. This gene is expressed at high levels during embryonic development; in adults, significant expression is found in Schwann cells, meningeal cells, lens and nerve. Mutations in this gene are associated with neurofibromatosis type II which is characterized by nervous system and skin tumors and ocular abnormalities. Two predominant isoforms and a number of minor isoforms are produced by alternatively spliced transcripts.
Specifications
| NF2 | |
| Polyclonal | |
| Unconjugated | |
| Nf2 | |
| ACN, BANF, Merlin, MGC123124, moesin-ezrin-radixin like, moesin-ezrin-radixin-like protein, moesin-ezrin-radizin-like protein, neurofibromatosis 2, neurofibromin 2, neurofibromin 2 (bilateral acoustic neuroma), neurofibromin 2 (merlin), neurofibromin 2b (merlin), neurofibromin-2, Nf2, Nf-2, NF2a, nf2b, OTTHUMP00000199607, SCH, Schwannomerlin, schwannomin | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 18016, 25744, 4771 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 3.36 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide, pH 7.3 | |
| P35240, P46662, Q63648 | |
| Nf2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 477-576 of human NF2 (NP_0002591) | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.