missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NF-M Rabbit anti-Human, Clone: 5E0D2, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | NF-M |
|---|---|
| Clone | 5E0D2 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18352392
|
Novus Biologicals
NBP3-15655-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18373715
|
Novus Biologicals
NBP3-15655-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NF-M Monoclonal antibody specifically detects NF-M in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| NF-M | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Autophagy, Cell Biology, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 4741 | |
| IgG | |
| Affinity purified |
| 5E0D2 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| NEF3neurofilament medium polypeptide, Neurofilament 3, neurofilament, medium polypeptide, neurofilament, medium polypeptide 150kDa, neurofilament-3 (150 kD medium), NF-M160 kDa neurofilament protein, NFMNeurofilament triplet M protein | |
| A synthetic peptide corresponding to a sequence within amino acids 600-700 of human NF-M (P07197). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel