missing translation for 'onlineSavingsMsg'
Learn More

NF-M Antibody (327921) [DyLight 594], Novus Biologicals™

Product Code. 30733801 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30733801 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733801 Supplier Novus Biologicals Supplier No. FAB3029M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NF-M Monoclonal antibody specifically detects NF-M in Human samples. It is validated for Western Blot,Gene Knock-Out
TRUSTED_SUSTAINABILITY

Specifications

Antigen NF-M
Applications Western Blot, Gene Knock-Out
Classification Monoclonal
Clone 327921
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias NEF3neurofilament medium polypeptide, Neurofilament 3, neurofilament, medium polypeptide, neurofilament, medium polypeptide 150kDa, neurofilament-3 (150 kD medium), NF-M160 kDa neurofilament protein, NFMNeurofilament triplet M protein
Host Species Mouse
Immunogen E. coli-derived recombinant human NF-M, Ser2-Ser358, Accession # P07197
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cell Biology, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 4741
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.