missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NF-L Rabbit anti-Human, Rat, Porcine, Bovine, Chicken, Clone: NEFL/2983R, Novus Biologicals™
Rabbit Monoclonal Antibody
£412.00
Specifications
| Antigen | NF-L |
|---|---|
| Clone | NEFL/2983R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 0.25-0.5 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
Description
NF-L Monoclonal specifically detects NF-L in Human, Rat, Porcine, Bovine, Chicken samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| NF-L | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Autophagy, Cell Biology, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience | |
| 10 mM PBS with 0.05% BSA | |
| 4747 | |
| IgG | |
| Protein A or G purified |
| NEFL/2983R | |
| Immunohistochemistry-Paraffin 0.25-0.5 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat, Pig, Bovine, Chicken | |
| 68 kDa neurofilament protein, neurofilament protein, light chain, neurofilament subunit NF-L, Neurofilament triplet L protein, neurofilament, light polypeptide, neurofilament-light, NF68FLJ53642, NF-L, NFLlight polypeptide 68kDa | |
| Recombinant full-length human NF-L protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title