missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NF-L Rabbit anti-Human, Rat, Porcine, Bovine, Chicken, Clone: NEFL/2983R, Novus Biologicals™
Shop All Bio Techne ProductsDescription
NF-L Monoclonal specifically detects NF-L in Human, Rat, Porcine, Bovine, Chicken samples. It is validated for Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | NF-L |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | NEFL/2983R |
| Concentration | 0.2 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry-Paraffin 0.25-0.5 ug/ml |
| Formulation | 10 mM PBS with 0.05% BSA |
| Gene Alias | 68 kDa neurofilament protein, neurofilament protein, light chain, neurofilament subunit NF-L, Neurofilament triplet L protein, neurofilament, light polypeptide, neurofilament-light, NF68FLJ53642, NF-L, NFLlight polypeptide 68kDa |
| Host Species | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?