Learn More
NF-L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12998-1-AP-20UL
 View more versions of this product
Product Code. 16801004
  • £135.00 / 20µL
Description
This antibody is specific to NEFL. It has a very weak reaction to NEFM.
Involved in the maintece of neuronal caliber, neurofilaments are the intermediate filament proteins found specifically in neurons, and are composed predomitly of three major proteins called NF-L, NF-M and NF-H. Like most other intermediate filament proteins (IFPs), the expression of the different neuronal IFPs is both tissue-specific and developmentally regulated. NF-L is the light or low molecular weight microfilament subunit and runs on SDS-PAGE gels at approximately 70 kDa. Neurofilament are the 10nm or intermediate filament proteins found specifically in neurons, and are composed predomitly of three major proteins called NF-L, NF-M and NF-H. NF-H is the heavy or high molecular weight microfilament subunit and runs on SDS-PAGE gels in the range 180-220kDa, with some variation in different species.
Specifications
Specifications
NF-L
Polyclonal
Unconjugated
NEFL
68 kDa neurofilament protein, CMT1F, CMT2E, NEFL, NF L, NF68, NFL, NF-L
Rabbit
Antigen Affinity Chromatography
RUO
18039, 4747, 83613
-20°C
Liquid
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
0.43 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P07196, P08551, P19527
Nefl
NF-L Fusion Protein Ag3667
20 μL
Primary
Rat, Human, Mouse