missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NF-L Antibody (SPM204), PE , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11576PE
This item is not returnable.
View return policy
Description
NF-L Monoclonal antibody specifically detects NF-L in Human, Rat, Porcine, Bovine, Chicken samples. It is validated for Flow Cytometry
Specifications
| NF-L | |
| Monoclonal | |
| PE | |
| 68 kDa neurofilament protein, neurofilament protein, light chain, neurofilament subunit NF-L, Neurofilament triplet L protein, neurofilament, light polypeptide, neurofilament-light, NF68FLJ53642, NF-L, NFLlight polypeptide 68kDa | |
| Neurofilament Purified from human brain | |
| 0.1 mL | |
| Autophagy, Cell Biology, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience | |
| 4747 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry | |
| SPM204 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat, Pig, Bovine, Chicken | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction