missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NF-L Antibody (CL4729) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59061JF669
This item is not returnable.
View return policy
Description
NF-L Monoclonal antibody specifically detects NF-L in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| NF-L | |
| Monoclonal | |
| Janelia Fluor 669 | |
| 68 kDa neurofilament protein, neurofilament protein, light chain, neurofilament subunit NF-L, Neurofilament triplet L protein, neurofilament, light polypeptide, neurofilament-light, NF68FLJ53642, NF-L, NFLlight polypeptide 68kDa | |
| This antibody was developed using a recombinant protein derived from P07196, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Autophagy, Cell Biology, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience | |
| 4747 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL4729 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction