missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ NF-kB II Phospho-Specific Array
Description
- This product must be accompanied with Antibody Array Detection Kit
- The arrays are printed on standard-size coated glass microscope slides and can be scanned on all microarray scanners that are compatible with 76 x 25 x 1 mm slides
- Suitable Sample: Cultured Cells, Frozen Tissue, FFPE Tissue, Serum
Specifications
Specifications
| Content And Storage | 4°C |
| Detection Method | Fluorometric |
| Quantity | 1 Kit (2 Samples) |
| For Use With (Application) | Profiling |
| Product Type | Array/Library |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?