Learn More
Invitrogen™ NF-H Monoclonal Antibody (RNF 405)

Mouse Monoclonal Antibody
Brand: Invitrogen™ OMA106117
Description
OMA1-06117 reacts exclusively with the phosphorylated form of the 200 kDa neurofilament from human, mouse, rat, canine, rabbit, monkey, bovine, ovine, hamster, chicken, guinea pig, and Xenopus laevis tissues. OMA1-06117 has been successfully used in Western blot and IHC (P) procedures. By Western blot, this antibody detects a single ~200 kDa protein representing phosphorylated neurofilament from rat brain homogenate. OMA1-06117 antigen is cytoskeletal preparation of calf brain tissue.
Involved in the maintenance of neuronal caliber, neurofilaments are the intermediate filament proteins found specifically in neurons, and are composed predominantly of three major proteins called NF-L, NF-M and NF-H. Like most other intermediate filament proteins (IFPs), the expression of the different neuronal IFPs is both tissue-specific and developmentally regulated. NF-H has an important role in mature axons and is involved in amyotrophic lateral sclerosis (ALS).
Specifications
| NF-H | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| P12036, P16884, P19246 | |
| Nefh, nefh.L | |
| Neurofilament preparation of calf brain tissue. | |
| 100 μg | |
| Primary | |
| Bovine, Canine, Chicken, Guinea Pig, Hamster, Human, Mouse, Monkey, Sheep, Rabbit, Rat, Rat, Zebrafish | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
| RNF 405 | |
| Unconjugated | |
| Nefh | |
| 200 kDa neurofilament protein; CMT2CC; heavy neurofilament protein; I79_012512; intermediate filament protein; KIAA0845; LOW QUALITY PROTEIN: neurofilament heavy polypeptide; mKIAA0845; NEF3; Nefh; nefh.L; neurofilament; neurofilament 200kDa; neurofilament heavy; neurofilament heavy L homeolog; neurofilament heavy polypeptide; neurofilament heavy polypeptide-like; neurofilament triplet H protein; neurofilament, heavy polypeptide; neurofilament, heavy polypeptide 200kDa; neurofilament, heavy polypeptide L homeolog; neurofilament-H; NF 200 kD; NF200; Nfh; NF-H; NHC; XELAEV_18007418mg | |
| Mouse | |
| Antigen affinity chromatography | |
| RUO | |
| 100101753, 100357177, 100716406, 100753127, 101107472, 101838568, 108715402, 24587, 380684, 417020, 442940, 4744, 528842 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.