missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NF-H Monoclonal Antibody (9B12)

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA547371

 View more versions of this product

Product Code. 17895087

  • £1482.00 / 500µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

NF-H Monoclonal Antibody for Western Blot, ICC/IF, IHC (PFA)

Involved in the maintenance of neuronal caliber, neurofilaments are the intermediate filament proteins found specifically in neurons, and are composed predominantly of three major proteins called NF-L, NF-M and NF-H. Like most other intermediate filament proteins (IFPs), the expression of the different neuronal IFPs is both tissue-specific and developmentally regulated. NF-H has an important role in mature axons and is involved in amyotrophic lateral sclerosis (ALS).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NF-H
Monoclonal
1 mg/mL
PBS with 50% glycerol and 5mM sodium azide
P12036, P16884, P19246
Nefh
Native NF-H purified from bovine spinal cord, binding to phosphorylated KSP sequences.
500 μL
Primary
Human, Mouse, Rat, Bovine
Antibody
IgG2b
Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry
9B12
Unconjugated
Nefh
200 kDa neurofilament protein; CMT2CC; heavy neurofilament protein; I79_012512; intermediate filament protein; KIAA0845; LOW QUALITY PROTEIN: neurofilament heavy polypeptide; mKIAA0845; NEF3; Nefh; nefh.L; neurofilament; neurofilament 200kDa; neurofilament heavy; neurofilament heavy L homeolog; neurofilament heavy polypeptide; neurofilament heavy polypeptide-like; neurofilament triplet H protein; neurofilament, heavy polypeptide; neurofilament, heavy polypeptide 200kDa; neurofilament, heavy polypeptide L homeolog; neurofilament-H; NF 200 kD; NF200; Nfh; NF-H; NHC; XELAEV_18007418mg
Mouse
Affinity chromatography
RUO
24587, 380684, 4744, 528842
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.