missing translation for 'onlineSavingsMsg'
Learn More

NF-H Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£143.00 - £375.00

Specifications

Antigen NF-H
Dilution Western Blot 1:10000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry Free-Floating 1:2000
Applications Immunohistochemistry (Free Floating), Immunocytochemistry, Immunofluorescence, ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18252703
View Documents
Novus Biologicals
NBP1-05215-0.025ML
0.025 mL
£143.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18238300
View Documents
Novus Biologicals
NBP1-05215
0.1 mL
£375.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NF-H Polyclonal specifically detects NF-H in Rat, Chicken samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry Free-Floating.
Spécification

Spécification

NF-H
Immunohistochemistry (Free Floating), Immunocytochemistry, Immunofluorescence, ELISA, Western Blot
Unconjugated
Rabbit
Cytoskeleton Markers, Neurodegeneration, Neurofilaments, Neuronal Cell Markers, Neuroscience
Whole antisera with No Preservative
4744
Purified rat Neurofilament Heavy construct containing most of the tandem KSP repeats [UniProt# P16884]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot 1:10000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry Free-Floating 1:2000
Polyclonal
Antisera
RUO
Rat, Chicken
200 kDa neurofilament protein, KIAA0845, Neurofilament Heavy (200kDa), neurofilament heavy polypeptide, Neurofilament triplet H protein, neurofilament, heavy polypeptide, neurofilament, heavy polypeptide 200kDa, NF200, NFH, NF-H
NEFH
IgG
Unpurified
200 kDa
Vidéos
FDS
Documents

Documents

Certificats

For Research Use Only

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis