missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
NF-H Monoclonal antibody specifically detects NF-H in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry Free-Floating
Specifications
Specifications
| Antigen | NF-H |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Free Floating) |
| Classification | Monoclonal |
| Clone | AH1 |
| Conjugate | Alexa Fluor 647 |
| Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry Free-Floating |
| Formulation | 50mM Sodium Borate |
| Gene Alias | 200 kDa neurofilament protein, KIAA0845, Neurofilament Heavy (200kDa), neurofilament heavy polypeptide, Neurofilament triplet H protein, neurofilament, heavy polypeptide, neurofilament, heavy polypeptide 200kDa, NF200, NFH, NF-H |
| Host Species | Mouse |
| Immunogen | Native axonal phosphorylated Neurofilament Heavy protein purified from bovine spinal cord |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?