missing translation for 'onlineSavingsMsg'
Learn More

NF-H Antibody (9B12), DyLight 594, Novus Biologicals™

Product Code. 18405115 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18405115 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18405115 Supplier Novus Biologicals Supplier No. NBP105210DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NF-H Monoclonal antibody specifically detects NF-H in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry Free-Floating
TRUSTED_SUSTAINABILITY

Specifications

Antigen NF-H
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Free Floating)
Classification Monoclonal
Clone 9B12
Conjugate DyLight 594
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry Free-Floating
Formulation 50mM Sodium Borate
Gene Alias 200 kDa neurofilament protein, KIAA0845, Neurofilament Heavy (200kDa), neurofilament heavy polypeptide, Neurofilament triplet H protein, neurofilament, heavy polypeptide, neurofilament, heavy polypeptide 200kDa, NF200, NFH, NF-H
Host Species Mouse
Immunogen Native NF-H purified from bovine spinal cord, binding to phosphorylated KSP sequences.
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cell Biology, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 4744
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.