Learn More
Tocris Bioscience™ NFκB p50 Transcription Factor Assay
GREENER_CHOICE
Product Code. 18798036
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. 18798036
Brand: Tocris Bioscience™ 7051/25
Includes
  • Transcription Factor Assay Plate
  • TFA Buffer, 5x
  • Blocking Reagent
  • Blocking Reagent
  • Primary Antibody
  • Gt X Rb, HRP Secondary Antibody
  • NFkB Capture Probe
  • NFkB Specific Competitor Oligonucleotide
  • TFA Negative Control Probe
  • HeLa Whole Cell Extract, TNFalpha Treated
  • Substrate (TMB/E)
  • Stop Solution
The transcription factor NFkB is involved in the expression and regulation of a number of important cellular and physiological processes such as growth, development, apoptosis, immune and inflammatory response
Tasquinimod is a high affinity negative allosteric modulator of HDAC4 (Kd = 10 - 30 nM). Binds the regulatory Zn2+ binding domain of HDAC4. Suppresses hypoxia-induced decrease in histone acetylation in human prostate cancer cells in vitro. Also binds S100A9. Antiangiogenic. Inhibits endothelial sprouting in vitro and growth of prostate tumor xenografts in nude mice. Orally bioavailable.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 25 mg