missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NF 023

Description
A subtype-selective, competitive and reversible P2X1 receptor antagonist. Displays IC50 values of 0.21, 28.9, > 50 and > 100 μM for human P2X1, P2X3, P2X2, and P2X4-mediated responses respectively. Selective over adrenoceptors, histamine and P2Y receptors. Also selectively inhibits the α-subunit of Go/i (EC50 ∽300 nM). Also inhibits DNA-binding activity of HMGA2 (IC50 = 10.63 μM).
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 1162.86 |
| Purity | >98% |
| Molecular Weight (g/mol) | 1162.851 |
| Chemical Name or Material | NF 023 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?