missing translation for 'onlineSavingsMsg'
Learn More

Nezukone, MedChemExpress

Product Code. 16483898
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16483898

Brand: Medchem Express HY103708

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Nezukone is a derivative of hinokitiol. Hinokitiol can restore iron transports, however, Nezukone cannot bind or transport iron.

Nezukone is a derivative of hinokitiol. Hinokitiol can restore iron transports, however, Nezukone cannot bind or transport iron.

Specifications

Chemical Name or Material Nezukone
Physical Form Liquid
CAS 13656-81-0
Color Brown
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C10H12O
Synonym C2-deoxyhinokitiol 4-Isopropyltropone
Solubility Information DMSO : ≥ 100 mg/mL (674.76 mM) ∣Ethanol : 100 mg/mL (674.76 mM; Need ultrasonic)
SMILES O=C1C=CC=C(C(C)C)C=C1
Molecular Weight (g/mol) 148.2
Formula Weight 148.2
Percent Purity 98.6%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.