Learn More
Nexilin Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27737-1-AP-150UL
Description
The Nexilin antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Nexilin. This antibody recognizes human, mouse, rat antigen. The Nexilin antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| Nexilin | |
| 650 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, Immunofluorescence, Immunohistochemistry, ELISA | |
| F actin binding protein, NELIN, Nexilin, NEXN | |
| 150 μL | |
| 91624 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |