missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Facilitate the scale-up of human pluripotent stem cells (hPSCs), creating a high-throughput platform for modeling various tissues and disease states.
- This ready-to-use hydrogel system features an optimized formulation that supports the rapid expansion of high-quality 3D stem cell spheroids with pluripotent characteristics.
- hPSCs can be directly thawed from liquid nitrogen or transferred from 2D matrix-coated culture vessels and immediately mixed with the hydrogel solution for static suspension cultures.
- The optimized protocol is particularly beneficial for time-sensitive experiments, as it minimizes the need for frequent medium exchanges, thereby saving time and resources.
- This hydrogel system is compatible with most hPSC culture media and tissue culture vessels.
- The unique static suspension culture method eliminates the need for microcarriers in large-scale bioreactors, simplifying and streamlining cell harvesting.
- The 3D stem cell spheroids generated using this system can be utilized for further sub-culturing, patterned differentiation, organoid development, or re-establishing 2D culture morphologies.
Specifications
Specifications
| Cell Type | hPSCs |
| Form | Liquid |
| Product Type | Hydrogel System |
| Quantity | 2 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?