missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Newcastle Disease Virus (NDV) Antibody (9F7) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB110-59912JF585
This item is not returnable.
View return policy
Description
Newcastle Disease Virus (NDV) Monoclonal antibody specifically detects Newcastle Disease Virus (NDV) in Virus samples. It is validated for Western Blot,Haemagglutination,Sandwich ELISA
Specifications
| Newcastle Disease Virus (NDV) | |
| Monoclonal | |
| Janelia Fluor 585 | |
| NDV | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Newcastle disease virus (La-sota strain). | |
| 0.2 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Hemagglutination Assay, Sandwich ELISA | |
| 9F7 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction