missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ New Zealand Bovine Thrombin, High Specific Activity

Product Code. p-7238941 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 KU
500 KU
Unit Size:
100ku
500ku
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17562896 100 KU 100ku
17572896 500 KU 500ku
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17562896 Supplier MP Biomedicals™ Supplier No. 0218053980
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Manufactured in a state-of-the-art facility in New Zealand using a chromatographic extraction process, under Quality Systems that are ISO Certified and audited to cGMP Principles.

Thrombin is used throughout the diagnostics industry in a variety of coagulation assays, clotting factor tests and defibrination of blood or plasma for serum controls. It can also be applied to site specific cleavage of recombinant fusion proteins and in biochemical and medical research applications, specifically in hemostasis applications.

This bovine thrombin with high specific activity is manufactured in New Zealand from bovine plasma of New Zealand origin using prothrombin activated with thromboplastin extracted from bovine lung tissue. Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

  • Lyophilized in glass vials under vacuum to optimize product performance
  • Specific activity of >2000 NIH units/mg protein

Specifications

Quantity 100 KU
Regulatory Status RUO
Species Bovine
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.