missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ New Zealand Bovine Thrombin, High Specific Activity

Product Code. 17562896 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 KU
500 KU
Unit Size:
100ku
500ku
This item is not returnable. View return policy

Product Code. 17562896

Brand: MP Biomedicals™ 0218053980

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Manufactured in a state-of-the-art facility in New Zealand using a chromatographic extraction process, under Quality Systems that are ISO Certified and audited to cGMP Principles.

Thrombin is used throughout the diagnostics industry in a variety of coagulation assays, clotting factor tests and defibrination of blood or plasma for serum controls. It can also be applied to site specific cleavage of recombinant fusion proteins and in biochemical and medical research applications, specifically in hemostasis applications.

This bovine thrombin with high specific activity is manufactured in New Zealand from bovine plasma of New Zealand origin using prothrombin activated with thromboplastin extracted from bovine lung tissue. Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

  • Lyophilized in glass vials under vacuum to optimize product performance
  • Specific activity of >2000 NIH units/mg protein
TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 KU
Regulatory Status Research Use Only
Species Bovine
Form Lyophilized
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt