Learn More
MP Biomedicals™ New Zealand Bovine Thrombin, High Specific Activity
Shop All MP Biomedicals ProductsDescription
Thrombin is used throughout the diagnostics industry in a variety of coagulation assays, clotting factor tests and defibrination of blood or plasma for serum controls. It can also be applied to site specific cleavage of recombinant fusion proteins and in biochemical and medical research applications, specifically in hemostasis applications.
This bovine thrombin with high specific activity is manufactured in New Zealand from bovine plasma of New Zealand origin using prothrombin activated with thromboplastin extracted from bovine lung tissue. Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
- Lyophilized in glass vials under vacuum to optimize product performance
- Specific activity of >2000 NIH units/mg protein
Specifications
Specifications
| Quantity | 500 KU |
| Regulatory Status | Research Use Only |
| Species | Bovine |
| Form | Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.